missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-hydroxypyridine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-2-hydroxypyridine |
| CAS | 13466-43-8 |
| Molecular Formula | C5 H4 Br N O |
| Quantity | 10 g |
| Synonym | 2(1H)-Pyridinone, 3-bromo-, 2(1H)-Pyridone, 3-bromo- (6CI,7CI,8CI), 3-Bromo-2(1H)-pyridinone, 3-Bromo-2(1H)-pyridone, 3-Bromo-2-hydroxypyridine, 3-Bromo-2-oxo-1,2-dihydropyridine, 3-Bromo-2-pyridinone, 3-Bromo-2-pyridone, 3-Bromopyridin-2-ol |
| InChI Formula | InChI=1S/C5H4BrNO/c6-4-2-1-3-7-5(4)8/h1-3H,(H,7,8) |
| SMILES | Oc1ncccc1Br |
| IUPAC Name | 3-bromopyridin-2-ol |
| Molecular Weight (g/mol) | 173.995 |
| Formula Weight | 172.948 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction