missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-hydroxypyridine, 97%, Thermo Scientific™

Product Code. 11916481 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11916481

Brand: Thermo Scientific Acros 446040010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 13466-43-8
Melting Point 182°C to 184°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C5H4BrNO
MDL Number MFCD03411569
Quantity 1 g
Synonym 3-bromo-2-hydroxypyridine, 3-bromopyridin-2-ol, 3-bromopyridin-2 1h-one, 3-bromo-2-pyridinol, 3-bromo-2 1h-pyridinone, 2 1h-pyridinone, 3-bromo, 2-hydroxy-3-bromopyridine, 3-bromo-2 1h-pyridone, bromopyridone, 3-bromopyridone
InChI Key YDUGVOUXNSWQSW-UHFFFAOYSA-N
SMILES BrC1=CC=CNC1=O
IUPAC Name 3-bromo-1H-pyridin-2-one
Molecular Weight (g/mol) 174.00
PubChem CID 818549
Formula Weight 174
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-hydroxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-2-hydroxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.