missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-hydroxybenzonitrile, 97%, Thermo Scientific™

Catalog Number p-7057068
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13073-28-4
C7H4BrNO
198.02
MFCD11042854
BUWXNBPLRLEXOZ-UHFFFAOYSA-N
2-bromo-6-cyanophenol, benzonitrile,3-bromo-2-hydroxy, acmc-1c4pd, 2-hydroxy-3-bromobenzonitrile, 3-bromo-2-hydroxy-benzonitrile, benzonitrile, 3-bromo-2-hydroxy, 3-bromo-2-hydroxybenznitrile, 3-bromo-2-hydroxybenzonitrile, -8-hydroxy-2-dipropylamino tetralin hydrobromide
12468680
3-bromo-2-hydroxybenzonitrile
OC1=C(C=CC=C1Br)C#N
This item is not returnable. View return policy
13073-28-4
C7H4BrNO
198.02
MFCD11042854
BUWXNBPLRLEXOZ-UHFFFAOYSA-N
2-bromo-6-cyanophenol, benzonitrile,3-bromo-2-hydroxy, acmc-1c4pd, 2-hydroxy-3-bromobenzonitrile, 3-bromo-2-hydroxy-benzonitrile, benzonitrile, 3-bromo-2-hydroxy, 3-bromo-2-hydroxybenznitrile, 3-bromo-2-hydroxybenzonitrile, -8-hydroxy-2-dipropylamino tetralin hydrobromide
12468680
3-bromo-2-hydroxybenzonitrile
OC1=C(C=CC=C1Br)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 118°C to 122°C
Color Brown to White
Quantity 1 g
UN Number UN3439
Formula Weight 198.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Bromo-2-hydroxybenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'