missing translation for 'onlineSavingsMsg'
Learn More

3-bromo-2-hydroxy-5-nitrobenzaldehyde, Thermo Scientific™

Product Code. 10148504
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10148504 5 g 5g
10507883 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10148504 Supplier Thermo Scientific Maybridge Supplier No. BTB03755DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16789-84-7
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.02
MDL Number MFCD00051833
InChI Key BESBCGANGAEHPM-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-nitrobenzaldehyde, 3-bromo-5-nitrosalicylaldehyde, 3-bromo-2-hydroxy-5-nitrobenzenecarbaldehyde, 2-hydroxy-3-bromo-5-nitrobenzaldehyde, 3-bromo-2-hydroxy-5-nitro-benzaldehyde, acmc-1cfaz, 3-bromo-5-nitro-salicylaldehyde, benzaldehyde,3-bromo-2-hydroxy-5-nitro, 3-bromanyl-5-nitro-2-oxidanyl-benzaldehyde
PubChem CID 519307
IUPAC Name 3-bromo-2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=C(Br)C=C(C=C1C=O)[N+]([O-])=O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 246.02
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-hydroxy-5-nitrobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.