missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-2-hydroxy-5-methylpyridine, 97%

Product Code. 11329845
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329845 1 g 1g
11339845 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329845 Supplier Alfa Aesar™ Supplier No. L19849.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17282-02-9
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD03791312
InChI Key KHBRMXVUQOVORD-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-methylpyridine, 3-bromo-5-methylpyridin-2-ol, 3-bromo-5-methylpyridin-2 1h-one, 3-bromo-2-hydroxy-5-picoline, 3-bromo-5-methyl-pyridin-2-ol, 3-bromo-5-methyl-2 1h-pyridinone, 3-bromo-5-methyl-2-pyridone, 3-bromo-5-methyl-2-pyridinol, 2 1h-pyridinone, 3-bromo-5-methyl, 3-bromo-2-hydroxy-5-picoline 3-bromo-2-hydroxy-5-methylpyridine
PubChem CID 3725261
IUPAC Name 3-bromo-5-methyl-1H-pyridin-2-one
SMILES CC1=CNC(=O)C(=C1)Br

Specifications

Melting Point 156°C to 158°C
Quantity 1 g
Beilstein 1423374
Formula Weight 188.02
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-hydroxy-5-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.