missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number 11459350
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
161957-56-8
C7H4BrFO2
219.009
MFCD00665763
UVKURTLVTLRSSM-UHFFFAOYSA-N
3-bromo-2-fluoro-benzoic acid, 3-bromo-2-fluorobenzoicacid, 2-fluoro-3-bromobenzoic acid, benzoic acid, 3-bromo-2-fluoro, 3-bromo-2-fluoro benzoic acid, buttpark 14\01-30, 3-bromo-2-fluorobenzenecarboxylic acid, pubchem1318, acmc-209dnk, intermediates-zcf02220
2736313
3-bromo-2-fluorobenzoic acid
C1=CC(=C(C(=C1)Br)F)C(=O)O
This item is not returnable. View return policy
161957-56-8
C7H4BrFO2
219.009
MFCD00665763
UVKURTLVTLRSSM-UHFFFAOYSA-N
3-bromo-2-fluoro-benzoic acid, 3-bromo-2-fluorobenzoicacid, 2-fluoro-3-bromobenzoic acid, benzoic acid, 3-bromo-2-fluoro, 3-bromo-2-fluoro benzoic acid, buttpark 14\01-30, 3-bromo-2-fluorobenzenecarboxylic acid, pubchem1318, acmc-209dnk, intermediates-zcf02220
2736313
3-bromo-2-fluorobenzoic acid
C1=CC(=C(C(=C1)Br)F)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 168°C to 170°C
Quantity 1 g
Formula Weight 219.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-fluorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'