missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-ethoxypyridine, 95%, Thermo Scientific Chemicals

Product Code. 11352789
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352789 1 g 1g
15413757 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352789 Supplier Thermo Scientific Alfa Aesar Supplier No. H27690.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57883-25-7 | Molecular Formula: C7H8BrNO | Molecular Weight: 202.051 g/mol

3-Bromo-2-ethoxypyridin is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-ethoxypyridin is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57883-25-7
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD00234309
InChI Key KLQBFCAXXZMOLZ-UHFFFAOYSA-N
PubChem CID 10987308
IUPAC Name 3-bromo-2-ethoxypyridine
SMILES CCOC1=C(C=CC=N1)Br

Specifications

Quantity 1 g
Synonym 2-ethoxy-3-bromopyridine, pyridine, 3-bromo-2-ethoxy, 3-bromanyl-2-ethoxy-pyridine, 3-bromo-2-ethoxy-pyridine, pubchem6233, bromo-2 ethoxypyridine, 3-bromo-2-ethoxypyridine, pyridine,3-bromo-2-ethoxy
Formula Weight 202.05
Percent Purity 95%
Chemical Name or Material 3-Bromo-2-ethoxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.