missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-dimethoxypyridine, 98%, Thermo Scientific™

Product Code. 11446450
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446450 25 g 25g
11436450 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446450 Supplier Thermo Scientific Alfa Aesar Supplier No. H27564.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13445-16-4
Molecular Formula C7H8BrNO2
Molecular Weight (g/mol) 218.05
MDL Number MFCD08669938
InChI Key UUTYQBCAEWGFQL-UHFFFAOYSA-N
Synonym pyridine, 3-bromo-2,6-dimethoxy, acmc-1bvd1, ksc495m9b, 2,6-dimethoxy-3-bromopyridine, 2,6-dimethoxyl-3-bromopyridine, 3-bromanyl-2,6-dimethoxy-pyridine, 3-bromo-2 pound not6-dimethoxypyridine
PubChem CID 642860
IUPAC Name 3-bromo-2,6-dimethoxypyridine
SMILES COC1=NC(=C(C=C1)Br)OC

Specifications

Quantity 25 g
Formula Weight 218.05
Percent Purity 98%
Chemical Name or Material 3-Bromo-2,6-dimethoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.