missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-trimethylsilyl-1-propyne, 98%

Product Code. 15440278
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440278 25 g 25g
15430278 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440278 Supplier Thermo Scientific Alfa Aesar Supplier No. H53451.14
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38002-45-8 | C6H11BrSi | 191.143 g/mol

3-Bromo-1-trimethylsilyl-1-propyne is used in the preparation of tetrahydroisoquinoline-3-carboxylic acid derivatives. It acts as a propargylating agent and used for the alkylation of dianion of beta-keto esters at gamma-carbon. It plays an important role in the synthesis of allenic alcohols and terminal conjugated enynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-1-trimethylsilyl-1-propyne is used in the preparation of tetrahydroisoquinoline-3-carboxylic acid derivatives. It acts as a propargylating agent and used for the alkylation of dianion of beta-keto esters at gamma-carbon. It plays an important role in the synthesis of allenic alcohols and terminal conjugated enynes.

Solubility
Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38002-45-8
Molecular Formula C6H11BrSi
Molecular Weight (g/mol) 191.143
MDL Number MFCD00134460
InChI Key GAPRPFRDVCCCHR-UHFFFAOYSA-N
Synonym 3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r
PubChem CID 642589
IUPAC Name 3-bromoprop-1-ynyl(trimethyl)silane
SMILES C[Si](C)(C)C#CCBr

Specifications

Boiling Point 44°C to 45°C (2mmHg)
Quantity 25 g
Flash Point 63°C (145°F)
Refractive Index 1.483
Density 1.17
Linear Formula BrCH2C≡CSi(CH3)3
Beilstein 2037650
Solubility Information Immiscible with water.
Formula Weight 191.14
Percent Purity 98%
Chemical Name or Material 3-Bromo-1-trimethylsilyl-1-propyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.