missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-propanol, 95%

Product Code. 11419897
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419897 5 g 5g
11429897 25 g 25g
11439897 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419897 Supplier Thermo Scientific Alfa Aesar Supplier No. B22857.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-18-9 | C3H7BrO | 138.99 g/mol

3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

Solubility
Miscible with water. Slightly miscible with ethanol and ether.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-18-9
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.99
MDL Number MFCD00002942
InChI Key RQFUZUMFPRMVDX-UHFFFAOYSA-N
Synonym 3-bromo-1-propanol, trimethylene bromohydrin, 1-propanol, 3-bromo, 3-bromopropanol, 3-hydroxypropyl bromide, 1-bromo-3-propanol, 3-bromo-propan-1-ol, ccris 5979, 1-bromo-3-hydroxypropane, unii-3t4axh68fh
PubChem CID 12308
IUPAC Name 3-bromopropan-1-ol
SMILES OCCCBr

Specifications

Density 1.59
Boiling Point 71°C to 73°C (11 mmHg)
Flash Point 93°C (199°F)
Linear Formula BrCH2CH2CH2OH
Refractive Index 1.4885
Quantity 5 g
Beilstein 969160
Solubility Information Miscible with water. Slightly miscible with ethanol and ether.
Formula Weight 138.99
Percent Purity ≥97%
Chemical Name or Material 3-Bromo-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.