missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-1-phenylpropane, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
5g
Specifications
Specifications
| CAS | 637-59-2 |
| Molecular Formula | C9H11Br |
| MDL Number | MFCD00000257 |
| Quantity | 100 g |
| Synonym | 3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl |
| InChI Key | XMZQWZJMTBCUFT-UHFFFAOYSA-N |
| SMILES | BrCCCC1=CC=CC=C1 |
| IUPAC Name | (3-bromopropyl)benzene |
| Molecular Weight (g/mol) | 199.09 |
| PubChem CID | 12503 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction