missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-benzofuran, 97%, Thermo Scientific™

Product Code. 10255792
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10255792

Brand: Thermo Scientific Maybridge CC27510DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 59214-70-9
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
InChI Key ICJNAOJPUTYWNV-UHFFFAOYSA-N
Synonym 3-bromobenzofuran, 3-bromobenzo b furan, benzofuran, 3-bromo, 3-bromo-1-benzo b furan, 3-bromobenzofurane, 3-bromo-benzo b furan, 3-bromanyl-1-benzofuran, ksc269c4b
PubChem CID 640589
IUPAC Name 3-bromo-1-benzofuran
SMILES C1=CC=C2C(=C1)C(=CO2)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 3-Bromo-1-benzofuran
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.