missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1,2-propanediol, 97%

Product Code. 10299153 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299153 100 mL Glass bottle 100mL
10286450 5 mL Glass bottle 5mL
10562801 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10299153 Supplier Thermo Scientific Acros Supplier No. 209851000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4704-77-2 | C3H7BrO2 | 154.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4704-77-2
Molecular Formula C3H7BrO2
Molecular Weight (g/mol) 154.99
MDL Number MFCD00004711
InChI Key SIBFQOUHOCRXDL-UHFFFAOYSA-N
PubChem CID 20827
ChEBI CHEBI:18719
IUPAC Name 3-bromopropane-1,2-diol
SMILES C(C(CBr)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.7700g/mL
Boiling Point 72.0°C to 75.0°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH(OH)CH2OH
Refractive Index 1.5165 to 1.5195
Quantity 100 mL
Beilstein 01, I, 248
Specific Gravity 1.77
Synonym 3-bromo-1,2-propanediol, monobromoglycerol, alpha-bromohydrin, 1,2-propanediol, 3-bromo, bromodeoxyglycerol, glycerol bromohydrin, 1-bromo-2,3-propanediol, 3-bromodeoxyglycerol, rs-3-bromo-1,2-propanediol, 1,2-propanediol,3-bromo
Formula Weight 154.99
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 3-Bromo-1, 2-propanediol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-1,2-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.