missing translation for 'onlineSavingsMsg'
Learn More

3-[Boc(methyl)aminomethyl]azetidine, 95%

Product Code. 15456778
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15456778

Brand: Thermo Scientific Alfa Aesar H60310.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1053655-53-0 | C10H20N2O2 | 200.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1053655-53-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD08061967
InChI Key IIRJNOLNCXOXOR-UHFFFAOYSA-N
Synonym tert-butyl azetidin-3-ylmethyl methyl carbamate, 3-n-boc-methylaminomethyl azetidine, tert-butyl n-azetidin-3-ylmethyl-n-methylcarbamate, 3-boc methyl aminomethyl azetidine, tert-butyl azetidin-3-ylmethyl, azetidin-3-ylmethyl neopentyl carbamate, tert-butylazetidin-3-ylmethyl methyl carbamate, azetidin-3-ylmethyl-methyl-carbamic acid tert-buty
PubChem CID 37818419
IUPAC Name tert-butyl N-(azetidin-3-ylmethyl)-N-methylcarbamate
SMILES CC(C)(C)OC(=O)N(C)CC1CNC1

Specifications

Quantity 250 mg
Formula Weight 200.28
Percent Purity 95%
Chemical Name or Material 3-[Boc(methyl)aminomethyl]azetidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.