missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-aminomethyl)pyridine, 97%, Thermo Scientific Chemicals

Product Code. 15484518
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484518 5 g 5g
15494518 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484518 Supplier Thermo Scientific Alfa Aesar Supplier No. H58183.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102297-41-6
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.26
MDL Number MFCD06411296
InChI Key PHQXGCNECRISKB-UHFFFAOYSA-N
Synonym n-boc-3-aminomethyl pyridine, tert-butyl pyridin-3-ylmethyl carbamate, n-boc-3-aminomethylpyridine, 3-boc-aminomethyl pyridine, tert-butyl n-pyridin-3-ylmethyl carbamate, carbamic acid, 3-pyridinylmethyl-, 1,1-dimethylethyl ester, 3-n-tert-butyloxycarbonyl amino methyl pyridine, pyridin-3-ylmethyl-carbamic acid tert-butyl ester, ambkkkkk570, tert-butyl pyridin-3-yl methylcarbamate
PubChem CID 13948280
IUPAC Name tert-butyl N-(pyridin-3-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1=CN=CC=C1

Specifications

Boiling Point 130°C
Refractive Index 1.508
UN Number UN1993
Quantity 5 g
Flash Point 22°C (72°F)
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 3-(Boc-aminomethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.