missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)piperidine, 97%, Thermo Scientific Chemicals

Product Code. 15429537
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429537 5 g 5g
15419537 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429537 Supplier Thermo Scientific Alfa Aesar Supplier No. H52788.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172603-05-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03839941
InChI Key WUOQXNWMYLFAHT-UHFFFAOYSA-N
Synonym 3-boc-aminopiperidine, tert-butyl piperidin-3-ylcarbamate, 3-n-boc-aminopiperidine, tert-butyl n-piperidin-3-yl carbamate, 3-boc-amino piperidine, 3-n-boc-amino piperidine, 3-tert-butoxycarbonylamino piperidine, 3-aminopiperidine, 3-boc protected, piperidin-3-yl-carbamic acid tert-butyl ester, tert-butyl n-3-piperidyl carbamate
PubChem CID 4233041
IUPAC Name tert-butyl N-piperidin-3-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1CCCNC1

Specifications

Quantity 5 g
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.