missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Boc-amino)azetidine, 98%
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Description
It is an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate in organic syntheses.
Solubility
Soluble in water. (5g/100ml)
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
It is an intermediate in organic syntheses.
Solubility
Soluble in water. (5g/100ml)
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Boiling Point | 268°C |
| Solubility Information | Soluble in water. (5g/100ml) |
| Density | 1.06 |
| Quantity | 500 mg |
| Flash Point | 116°C (241°F) |
| Formula Weight | 172.23 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-(Boc-amino)azetidine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction