missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)azetidine, 98%

Product Code. 15455918
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
This item is not returnable. View return policy

Product Code. 15455918

Brand: Thermo Scientific Alfa Aesar H59556.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91188-13-5 | C8H16N2O2 | 172.23 g/mol

It is an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses.

Solubility
Soluble in water. (5g/100ml)

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91188-13-5
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.23
MDL Number MFCD01861759
InChI Key NEMXVXVJGXZDRR-UHFFFAOYSA-N
Synonym tert-butyl azetidin-3-ylcarbamate, 3-n-boc-amino-azetidine, 3-boc-aminoazetidine, 3-n-boc-amino azetidine, tert-butyl n-azetidin-3-yl carbamate, 3-n-boc-aminoazetidine, 3-boc-amino azetidine, azetidin-3-yl-carbamic acid tert-butyl ester, 3-boc-amino-azetidine, 3-tert-butoxycarbonylamino azetidine
PubChem CID 1519405
IUPAC Name tert-butyl N-(azetidin-3-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1CNC1

Specifications

Boiling Point 268°C
Solubility Information Soluble in water. (5g/100ml)
Density 1.06
Quantity 250 mg
Flash Point 116°C (241°F)
Formula Weight 172.23
Percent Purity 98%
Chemical Name or Material 3-(Boc-amino)azetidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.