missing translation for 'onlineSavingsMsg'
Learn More

3-Biphenylcarboxylic acid, 97%

Product Code. 10302963 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10302963 1 g Glass bottle 1g
10651935 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10302963 Supplier Thermo Scientific Acros Supplier No. 371770010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 716-76-7 | C13H10O2 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 716-76-7
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00045846
InChI Key XNLWJFYYOIRPIO-UHFFFAOYSA-N
Synonym biphenyl-3-carboxylic acid, 3-biphenylcarboxylic acid, 1,1'-biphenyl-3-carboxylic acid, diphenyl-3-carboxylic acid, wln: qvr cr, 3-biphenylcarboxylicacid, m-phenylbenzoic acid, 3-phenyl benzoic acid
PubChem CID 12854
IUPAC Name 3-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 164°C to 169°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5C6H4COOH
Quantity 1 g
Beilstein 09, IV, 2478
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone,diethyl ether,hexane,soluble in benzene,petroleum ether,alkali
Formula Weight 198.22
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Biphenylcarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.