missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Beta,5-Beta)-3-Hydroxycholan-24-oic acid, TRC
Specifications
Specifications
| Chemical Name or Material | Isolithocholic acid |
| CAS | 1534-35-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H40 O3 |
| Quantity | 250 mg |
| Synonym | (3β,5β)-3-Hydroxycholan-24-oic acid, 3-Epilithocholic acid, 3β-Hydroxy-5β-cholan-24-oic acid, 3β-Hydroxy-5β-cholanic acid, 3β-Hydroxy-5β-cholanoic acid, 3β-Lithocholic acid, 5β-Cholanic acid-3β-ol, Isolithocholic acid, β-Lithocholanic acid, β-Lithocholic acid |
| InChI Formula | InChI=1S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25H,4-14H2,1-3H3,(H,26,27)/t15-,16-,17+,18+,19-,20+,21+,23+,24-/m1/s1 |
| SMILES | C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (4R)-4-[(3S,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
| Molecular Weight (g/mol) | 376.57 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?