missing translation for 'onlineSavingsMsg'
Learn More

(3-Benzyloxypropyl)triphenylphosphonium Bromide, TRC

Product Code. 30345894
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30345894 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345894 Supplier Toronto Research Chemicals Supplier No. TRCB2879801G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3-Benzyloxypropyl)triphenylphosphonium Bromide
CAS 54314-85-1
Molecular Formula C28H28BrOP
Quantity 1 g
Synonym Triphenyl[3-(phenylmethoxy)propyl]-phosphonium Bromide
InChI Formula InChI=1S/C28H28OP.BrH/c1-5-14-25(15-6-1)24-29-22-13-23-30(26-16-7-2-8-17-26,27-18-9-3-10-19-27)28-20-11-4-12-21-28;/h1-12,14-21H,13,22-24H2;1H/q+1;/p-1
SMILES [Br-].C(COCc1ccccc1)C[P+](c2ccccc2)(c3ccccc3)c4ccccc4
IUPAC Name triphenyl(3-phenylmethoxypropyl)phosphanium;bromide
Molecular Weight (g/mol) 491.4
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.