missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Benzyloxycarbonylamino-5-nitrobenzeneboronic acid, 98%

Product Code. 15479885
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479885 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15479885 Supplier Alfa Aesar™ Supplier No. H52536.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 874219-56-4
Molecular Formula C14H13BN2O6
Molecular Weight (g/mol) 316.08
MDL Number MFCD08235090
InChI Key DNBCLFIDIDSVDL-UHFFFAOYSA-N
Synonym 3-benzyloxycarbonylamino-5-nitrophenylboronic acid, 3-benzyloxycarbonylamino-5-nitrophenyl boronic acid, 3-benzyloxy carbonyl amino-5-nitrophenyl boronic acid, 3-benzyloxy carbonyl amino-5-nitrophenylboronic acid, acmc-209qlh, 3-benzyloxycarbonylamino-5-nitrobenzeneboronic acid, 3-amino-5-nitrobenzeneboronic acid, cbz protected, 3-nitro-5-phenylmethoxycarbonylamino phenyl boronic acid, carbamic acid, 3-borono-5-nitrophenyl-,c-phenylmethyl ester 9ci
PubChem CID 44119858
IUPAC Name [3-nitro-5-(phenylmethoxycarbonylamino)phenyl]boronic acid
SMILES OB(O)C1=CC(NC(=O)OCC2=CC=CC=C2)=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 218°C to 220°C
Quantity 1 g
Formula Weight 316.08
Percent Purity 98%
Chemical Name or Material 3-Benzyloxycarbonylamino-5-nitrobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.