missing translation for 'onlineSavingsMsg'
Learn More

3-[(Benzyloxycarbonyl)amino]propionaldehyde, 95%

Product Code. 10471810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10471810 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10471810 Supplier Thermo Scientific Acros Supplier No. 427720010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65564-05-8 | C11H13NO3 | 207.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 65564-05-8
CAS Min % 94.0
CAS Max % 100.0
Melting Point 52°C to 57°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C11H13NO3
Linear Formula C6H5CH2OC(O)NHCH2CH2CHO
MDL Number MFCD05664298
Quantity 1 g
Synonym benzyl 3-oxopropylcarbamate, 3-benzyloxycarbonyl amino-1-propanal, 3-benzyloxycarbonyl amino propionaldehyde, benzyl n-3-oxopropyl carbamate, benzyl 3-oxopropyl carbamate, 3-oxo-propyl-carbamic acid benzyl ester, n-phenylmethoxycarbonyl-3-aminopropanal, n-cbz-3-aminopropanal, pubchem13633
InChI Key PQMOZOQTXKMYSK-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)COC(=O)NCCC=O
IUPAC Name benzyl N-(3-oxopropyl)carbamate
Molecular Weight (g/mol) 207.23
PubChem CID 10398106
Formula Weight 207.23
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-[(Benzyloxycarbonyl)amino]propionaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3-[(Benzyloxycarbonyl)amino]propionaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.