missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybromobenzene, 96%

Product Code. 10577980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10577980 50 g 50g
10280020 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10577980 Supplier Thermo Scientific Acros Supplier No. 437790500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53087-13-1 | C13H11BrO | 263.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53087-13-1
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.13
InChI Key HVWZMGZBJCJDOX-UHFFFAOYSA-N
Synonym 1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065
PubChem CID 2756638
IUPAC Name 1-bromo-3-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC(=CC=C2)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 60°C to 62°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Quantity 50 g
Formula Weight 263.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Benzyloxybromobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Benzyloxybromobenzene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.