missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzoic acid, 98%

Product Code. 15405647
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405647 1 g 1g
15415647 5 g 5g
15425647 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15405647 Supplier Thermo Scientific Alfa Aesar Supplier No. H32882.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 69026-14-8 | C14H11O3 | 227.24 g/mol

3-Benzyloxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzyloxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69026-14-8
Molecular Formula C14H11O3
Molecular Weight (g/mol) 227.24
MDL Number MFCD00527901
InChI Key CISXCTKEQYOZAM-UHFFFAOYSA-M
Synonym 3-benzyloxy benzoic acid, 3-benzyloxybenzoic acid, benzoic acid, 3-phenylmethoxy, 3-benzyloxy-benzoic acid, 3-phenylmethoxy benzoic acid, enamine_004835, acmc-1b5k3
PubChem CID 309226
IUPAC Name 3-phenylmethoxybenzoic acid
SMILES [O-]C(=O)C1=CC=CC(OCC2=CC=CC=C2)=C1

Specifications

Melting Point 133°C to 135°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 3-Benzyloxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.