missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzaldehyde, 98%

Product Code. 11459957
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459957 5 g 5g
11469957 25 g 25g
11479957 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459957 Supplier Thermo Scientific Alfa Aesar Supplier No. B22909.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1700-37-4 | C14H12O2 | 212.248 g/mol

3-Benzyloxybenzaldehyde is used as a pharmaceutical intermediate. It activates the adenyl cyclase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzyloxybenzaldehyde is used as a pharmaceutical intermediate. It activates the adenyl cyclase.

Solubility
Soluble in water 38.87 mg/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1700-37-4
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00003367
InChI Key JAICGBJIBWDEIZ-UHFFFAOYSA-N
Synonym 3-benzyloxybenzaldehyde, 3-benzyloxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, m-benzyloxybenzaldehyde, benzaldehyde, m-benzyloxy, 3-phenylmethoxy benzaldehyde, 3-benzyloxy-benzaldehyde, unii-k1s6ob2gnn, k1s6ob2gnn
PubChem CID 74342
IUPAC Name 3-phenylmethoxybenzaldehyde
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O

Specifications

Melting Point 54°C to 58°C
Boiling Point 180°C (0.08 mmHg)
Quantity 5 g
Beilstein 1956589
Sensitivity Air Sensitive
Solubility Information Soluble in water 38.87mg/L @ 25°C.
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 3-Benzyloxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.