missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzaldehyde, 98%, Thermo Scientific™

Product Code. 10132420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132420 100 g Glass bottle 100g
10761545 5 g Glass bottle 5g
10122420 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10132420 Supplier Thermo Scientific Acros Supplier No. 106071000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1700-37-4
MDL Number MFCD00003367
InChI Key JAICGBJIBWDEIZ-UHFFFAOYSA-N
Synonym 3-benzyloxybenzaldehyde, 3-benzyloxy benzaldehyde, benzaldehyde, 3-phenylmethoxy, m-benzyloxybenzaldehyde, benzaldehyde, m-benzyloxy, 3-phenylmethoxy benzaldehyde, 3-benzyloxy-benzaldehyde, unii-k1s6ob2gnn, k1s6ob2gnn
PubChem CID 74342
IUPAC Name 3-phenylmethoxybenzaldehyde
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55°C to 59°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C14H12O2
Linear Formula C6H5CH2OC6H4CHO
Quantity 100 g
Molecular Weight (g/mol) 212.25
Formula Weight 212.25
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Benzyloxybenzaldehyde, 98%
Show More Show Less

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.