missing translation for 'onlineSavingsMsg'
Learn More

3'-Benzyloxyacetophenone, 97%

Product Code. 11496087
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496087 100 g 100g
11476087 5 g 5g
11486087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496087 Supplier Thermo Scientific Alfa Aesar Supplier No. B25084.22
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34068-01-4 | C15H14O2 | 226.275 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34068-01-4
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00026221
InChI Key FGQMEAWGAUALJQ-UHFFFAOYSA-N
Synonym 1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene
PubChem CID 98689
IUPAC Name 1-(3-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2

Specifications

Quantity 100 g
Formula Weight 226.28
Percent Purity 97%
Chemical Name or Material 3'-Benzyloxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.