missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzylamino)propylamine, 95%, Thermo Scientific™

Product Code. 15492589
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13910-48-0
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.25
MDL Number MFCD00274361
InChI Key RFLHDXQRFPJPRR-UHFFFAOYSA-N
Synonym n1-benzylpropane-1,3-diamine, 3-benzylamino propylamine, n-benzylpropane-1,3-diamine, 3-aminopropyl benzyl amine, n-benzyl-1,3-propanediamine, n-3-aminopropyl-n-benzylamine, 3-aminopropyl benzylamine, n-benzylpropan-1,3-diamin, n-benzyltrimethylenediamine, n-3-aminopropyl benzylamine
PubChem CID 83811
IUPAC Name N'-benzylpropane-1,3-diamine
SMILES NCCCNCC1=CC=CC=C1

Specifications

Color Yellow
Refractive Index 1.532
Quantity 250 mg
Beilstein 509316
Formula Weight 164.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-(Benzylamino)propylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'