missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzylamino)propionitrile, 97+%

Product Code. 11314937
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314937 100 g 100g
11394927 5 g 5g
11304937 25 g 25g
11324937 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11314937 Supplier Thermo Scientific Alfa Aesar Supplier No. 041949.22
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 706-03-6 | C10H13N2 | 161.23 g/mol

3-(Benzyl amino)propionitrile was used as starting reagent in the synthesis of (3R,4S)-1-benzyl-4-phenylpyrrolidine-3-carboxylic acid, 3-[benzyl-(3,5-dimethyl-pyrazol-1-ylmethyl)-amino]-propionitrile. It undergoes aza-type Michael reaction with 1-hydroxymethyl-5-methyl-1H-pyrazole-3-carboxylic acid ethyl ester under mild conditions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 706-03-6
Molecular Formula C10H13N2
Molecular Weight (g/mol) 161.23
MDL Number MFCD00001955
InChI Key MWTGBAURSCEGSL-UHFFFAOYSA-O
Synonym 3-benzylamino propanenitrile, 3-benzylamino propionitrile, n-benzyl-2-cyanoethylamine, propanenitrile, 3-phenylmethyl amino, a-cyanoethyl benzylamine, beta-cyanoethylbenzylamine, acmc-1bk6i, 3-benzylamino-propionitrile
PubChem CID 69719
IUPAC Name 3-(benzylamino)propanenitrile
SMILES N#CCC[NH2+]CC1=CC=CC=C1

Specifications

Density 1.024 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.5308
Quantity 100 g
UN Number UN3276
Solubility Information Soluble in DMSO.
Formula Weight 160.22
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 3-(Benzylamino)propionitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.