missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylpyridine, 97%, Thermo Scientific™

Catalog Number p-63727
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5424-19-1
183.21
MFCD00006394
RYMBAPVTUHZCNF-UHFFFAOYSA-N
3-benzoylpyridine, phenyl pyridin-3-yl methanone, phenyl 3-pyridyl ketone, methanone, phenyl-3-pyridinyl, pyridine, 3-benzoyl, 3-pyridyl phenyl ketone, ketone, phenyl 3-pyridyl, 3-benzoyl pyridine, phenyl-b-pyridyl ketone, phenyl 3-pyridinyl methanone
21540
phenyl(pyridin-3-yl)methanone
C1=CC=C(C=C1)C(=O)C2=CN=CC=C2
This item is not returnable. View return policy
5424-19-1
183.21
MFCD00006394
RYMBAPVTUHZCNF-UHFFFAOYSA-N
3-benzoylpyridine, phenyl pyridin-3-yl methanone, phenyl 3-pyridyl ketone, methanone, phenyl-3-pyridinyl, pyridine, 3-benzoyl, 3-pyridyl phenyl ketone, ketone, phenyl 3-pyridyl, 3-benzoyl pyridine, phenyl-b-pyridyl ketone, phenyl 3-pyridinyl methanone
21540
phenyl(pyridin-3-yl)methanone
C1=CC=C(C=C1)C(=O)C2=CN=CC=C2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 36.0°C to 40.0°C
Color White to Yellow
Boiling Point 307.0°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H9NO
Linear Formula C6H5CO(C5H4N)
Quantity 5 g
Formula Weight 183.21
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 3-Benzoylpyridine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Benzoylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'