missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylpyridine, 97%, Thermo Scientific™

Product Code. 10444735 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444735 5 g Glass bottle 5g
10559800 25 g Glass bottle 25g
10458050 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10444735 Supplier Thermo Scientific Acros Supplier No. 105810050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5424-19-1
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006394
InChI Key RYMBAPVTUHZCNF-UHFFFAOYSA-N
Synonym 3-benzoylpyridine, phenyl pyridin-3-yl methanone, phenyl 3-pyridyl ketone, methanone, phenyl-3-pyridinyl, pyridine, 3-benzoyl, 3-pyridyl phenyl ketone, ketone, phenyl 3-pyridyl, 3-benzoyl pyridine, phenyl-b-pyridyl ketone, phenyl 3-pyridinyl methanone
PubChem CID 21540
IUPAC Name phenyl(pyridin-3-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CN=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 36.0°C to 40.0°C
Color White to Yellow
Boiling Point 307.0°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H9NO
Linear Formula C6H5CO(C5H4N)
Quantity 5 g
Formula Weight 183.21
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 3-Benzoylpyridine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Benzoylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.