missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylpropanoic Acid, TRC

Product Code. 30343204
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343204 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343204 Supplier Toronto Research Chemicals Supplier No. TRCB20880050G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Benzoylpropanoic Acid
CAS 2051-95-8
Molecular Formula C10 H10 O3
Quantity 50 g
Synonym Benzenebutanoic acid, γ-oxo-, Propionic acid, 3-benzoyl- (6CI,7CI,8CI), γ-Oxobenzenebutanoic acid, 3-Benzoylpropanoic acid, 3-Benzoylpropionic acid, 4-Oxo-4-phenylbutanoic acid, 4-Oxo-4-phenylbutyric acid, 4-Phenyl-4-oxobutanoic acid, NSC 2092, NSC 229040, NSC 51010, Propanoic acid, 3-benzoyl-, β-Benzoylpropionic acid
InChI Formula InChI=1S/C10H10O3/c11-9(6-7-10(12)13)8-4-2-1-3-5-8/h1-5H,6-7H2,(H,12,13)
SMILES OC(=O)CCC(=O)c1ccccc1
IUPAC Name 4-oxo-4-phenylbutanoic acid
Molecular Weight (g/mol) 178.185
Formula Weight 178.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.