missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3'-Azido-3'-deoxythymidine, 98%

Product Code. p-7043026
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15496739

Brand: Thermo Scientific Chemicals J65127.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anti-viral agent | CAS: 30516-87-1 | C10H13N5O4 | 267.25 g/mol

3'-Azido-3'-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3?-Azido-3?-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Azido-3′-deoxythymidine is a Anti-viral agent, and also used as a reverse transcriptase inhibitor active against HIV-1 virus that blocks the incorporation of nucleotides into growing DNA strands. Useful for antiviral and anticancer studies. 3′-Azido-3′-deoxythymidine enters CNS by diffusion and causes irreversible telomere shortening.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30516-87-1
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00006536
InChI Key HBOMLICNUCNMMY-CFQLRCIDNA-N
Synonym azt; azidothymidine; zdv; zidovudine, 1-2r-4-azido-5-hydroxymethyl oxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
PubChem CID 455007
SMILES CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O

Specifications

Melting Point 113°C to 115°C
Color White
Quantity 1 g
Beilstein 3595791
Sensitivity Light sensitive; Very Hygroscopic
Merck Index 1410123
Solubility Information Soluble in water.
IUPAC Name 1-[(2R,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
Formula Weight 267.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3'-Azido-3'-deoxythymidine
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.