missing translation for 'onlineSavingsMsg'
Learn More

3-Azido-1-propylamine

Product Code. p-7043129
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417029 100 mg 100mg
15407029 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417029 Supplier Thermo Scientific Alfa Aesar Supplier No. J65668.MC
Add to basket
This item is not returnable. View return policy

Azido-containing reagent with carbonyl and carboxyl reactivity | CAS: 88192-19-2 | C3H8N4 | 100.125 g/mol

Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.

Solubility
Soluble in DMSO, DMF, DCM, chloroform, THF, and water.Miscible with dimethylsulfoxide, dimethyl formamide, dichloromethane, tetrahydrofuran, chloroform and water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88192-19-2
Molecular Formula C3H8N4
Molecular Weight (g/mol) 100.125
MDL Number MFCD11046568
InChI Key OYBOVXXFJYJYPC-UHFFFAOYSA-N
Synonym 3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine
PubChem CID 150110
IUPAC Name 3-azidopropan-1-amine
SMILES C(CN)CN=[N+]=[N-]

Specifications

Color Yellow
Flash Point 60°C (140°F)
Linear Formula N3CH2CH2CH2NH2
Quantity 100 mg
UN Number UN1993
Solubility Information Soluble in DMSO,DMF,DCM,chloroform,THF,and water;Miscible with dimethylsulfoxide,dimethyl formamide,dichloromethane,tetrahydrofuran,chloroform and water.
Formula Weight 100.12
Physical Form Liquid
Chemical Name or Material 3-Azido-1-propylamine
compliance-icons compliance-icons
Product Identifier
  • 3-Azido-1-propylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Azido-1-propylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.