missing translation for 'onlineSavingsMsg'
Learn More

3-Azabicyclo[3.3.0]octane hydrochloride, 99%, Thermo Scientific™

Product Code. 15585986 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15585986 100 g 100g
15565986 5 g 5g
15575986 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15585986 Supplier Thermo Scientific Acros Supplier No. 457701000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 112626-50-3
Molecular Formula C7H13N·ClH
Molecular Weight (g/mol) 147.65
InChI Key HVZRRRPCVOJOLJ-UHFFFAOYSA-N
Synonym 3-azabicyclo 3.3.0 octane hydrochloride, octahydrocyclopenta c pyrrole hydrochloride, 3-azabicyclo 3.3.0 octane hcl, 3-azabicyclo 3,3,0 octane hcl, 3-azabicyclo 3.3.0 octanehydrochloride, 1,2,3,3a,4,5,6,6a-octahydrocyclopenta c pyrrole hydrochloride, pubchem9779, cis-octahydro-cyclopenta c pyrrole hydrochloride, c7h14ncl, 3-azabicyclo 3.3.0 octane hydrochloride;
PubChem CID 21572123
IUPAC Name 1,2,3,3a,4,5,6,6a-octahydrocyclopenta[c]pyrrole;hydrochloride
SMILES C1CC2CNCC2C1.Cl

Specifications

Melting Point 116.0°C to 118.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5 to max. 101.5% min. (Argentometry)
Quantity 100 g
Formula Weight 147.65
Percent Purity 99%
Chemical Name or Material 3-Azabicyclo[3.3.0]octane hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.