missing translation for 'onlineSavingsMsg'
Learn More

3-Aminothiophenol, 98%, Thermo Scientific™

Catalog Number 10021970
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 22948-02-3
CAS Min % 97.5
CAS Max % 100.0
Density 1.1790g/mL
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C6H6NS
Linear Formula H2NC6H4SH
Refractive Index 1.6560 to 1.6580
MDL Number MFCD00007791
Quantity 5g
Beilstein 13, 425
Specific Gravity 1.179
Synonym 3-aminothiophenol, benzenethiol, 3-amino, 3-mercaptoaniline, m-amino thiophenol, 3-aminobenzene-1-thiol, 3-amino thiophenol, 3-amino-benzenethiol, pubchem2470, acmc-1co9c
InChI Key KFFUEVDMVNIOHA-UHFFFAOYSA-M
SMILES NC1=CC=CC([S-])=C1
IUPAC Name (3-aminophenyl)sulfanide
Molecular Weight (g/mol) 124.18
PubChem CID 31577
Formula Weight 125.19
Percent Purity 98%
Chemical Name or Material 3-Aminothiophenol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminothiophenol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'