missing translation for 'onlineSavingsMsg'
Learn More

3-Aminothiophenol, 97%

Product Code. 11487146
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487146 5 g 5g
11497146 25 g 25g
11329519 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487146 Supplier Thermo Scientific Alfa Aesar Supplier No. B25473.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22948-02-3 | C6H6NS | 124.18 g/mol

3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22948-02-3
Molecular Formula C6H6NS
Molecular Weight (g/mol) 124.18
MDL Number MFCD00007791
InChI Key KFFUEVDMVNIOHA-UHFFFAOYSA-M
Synonym 3-aminothiophenol, benzenethiol, 3-amino, 3-mercaptoaniline, m-amino thiophenol, 3-aminobenzene-1-thiol, 3-amino thiophenol, 3-amino-benzenethiol, pubchem2470, acmc-1co9c
PubChem CID 31577
SMILES NC1=CC=CC([S-])=C1

Specifications

Density 1.179
Boiling Point 123°C to 125°C (8 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.659
Quantity 5 g
UN Number UN3267
Sensitivity Air sensitive
Solubility Information Soluble in water.
IUPAC Name 3-aminobenzenethiol
Formula Weight 125.19
Percent Purity 97%
Chemical Name or Material 3-Aminothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.