missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoquinuclidine Dihydrochloride, TRC

Product Code. 30311909
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311909 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311909 Supplier Toronto Research Chemicals Supplier No. TRCA6291851G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Aminoquinuclidine Dihydrochloride
CAS 6530-09-2
Molecular Formula C7H16Cl2N2
Quantity 1 g
Synonym 1-Azabicyclo[2.2.2]octan-3-amine Hydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane Dihydrochloride, NSC 93906
InChI Formula InChI=1S/C7H14N2.2ClH/c8-7-5-9-3-1-6(7)2-4-9;;/h6-7H,1-5,8H2;2*1H
SMILES Cl.Cl.NC1CN2CCC1CC2
IUPAC Name 1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
Molecular Weight (g/mol) 199.12
Formula Weight 198.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.