missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminoquinoline, TRC
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Specifications
Specifications
| Chemical Name or Material | 3-Aminoquinoline |
| CAS | 580-17-6 |
| Molecular Formula | C9H8N2 |
| Quantity | 100 mg |
| Synonym | 3-Quinolinamine (9CI, ACI), Quinoline, 3-amino- (8CI), (Quinolin-3-yl)amine, 3-Aminoquinoline, 3-Quinolylamine, NSC 7934, Quinolin-3-amine |
| InChI Formula | InChI=1S/C9H8N2/c10-8-5-7-3-1-2-4-9(7)11-6-8/h1-6H,10H2 |
| SMILES | Nc1cnc2ccccc2c1 |
| IUPAC Name | quinolin-3-amine |
| Molecular Weight (g/mol) | 144.17 |
| Formula Weight | 144.0687 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction