missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoquinoline, 99%

Product Code. 10082780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082780 5 g Glass bottle 5g
10616914 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082780 Supplier Thermo Scientific Acros Supplier No. 213780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 580-17-6 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-17-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006772
InChI Key SVNCRRZKBNSMIV-UHFFFAOYSA-N
Synonym 3-aminoquinoline, 3-quinolinamine, 3-quinolylamine, 3-quinolineamine, quinoline, 3-amino, quinolin-3-ylamine, 3-amino quinoline, unii-98ovl5k3r4, quinolin-3-yl-amine, ccris 1678
PubChem CID 11375
IUPAC Name quinolin-3-amine
SMILES C1=CC=C2C(=C1)C=C(C=N2)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 91.0°C to 94.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 22, I, 638
Solubility Information Solubility in water: slightly soluble in hot water. Other solubilities: soluble in methanol
Formula Weight 144.18
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Aminoquinoline, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.