missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoquinoline, 98%

Product Code. 11497927
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497927 5 g 5g
11487927 1 g 1g
11407937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497927 Supplier Thermo Scientific Alfa Aesar Supplier No. A16592.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 580-17-6 | C9H8N2 | 144.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-17-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006772
InChI Key SVNCRRZKBNSMIV-UHFFFAOYSA-N
Synonym 3-aminoquinoline, 3-quinolinamine, 3-quinolylamine, 3-quinolineamine, quinoline, 3-amino, quinolin-3-ylamine, 3-amino quinoline, unii-98ovl5k3r4, quinolin-3-yl-amine, ccris 1678
PubChem CID 11375
IUPAC Name quinolin-3-amine
SMILES C1=CC=C2C(=C1)C=C(C=N2)N

Specifications

Melting Point 92°C to 96°C
Quantity 5 g
Beilstein 113317
Sensitivity Air Sensitive
Formula Weight 144.18
Percent Purity 98%
Chemical Name or Material 3-Aminoquinoline
compliance-icons
Product Identifier
  • 3-Aminoquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Quinolinamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.