missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopyrazole, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3-Aminopyrazole |
| CAS | 1820-80-0 |
| Molecular Formula | C3 H5 N3 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C3H5N3/c4-3-1-2-5-6-3/h1-2H,(H3,4,5,6) |
| SMILES | Nc1cc[nH]n1 |
| IUPAC Name | 1H-pyrazol-3-amine |
| Molecular Weight (g/mol) | 83.09 |
| Formula Weight | 83.0483 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction