missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazole, 98%, Thermo Scientific Chemicals

Catalog Number p-3756833
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1820-80-0
C3H5N3
83.09
MFCD00005236
JVVRJMXHNUAPHW-UHFFFAOYSA-N
3-aminopyrazole, 1h-pyrazol-3-amine, 1h-pyrazole-5-amine, 5-aminopyrazole, 1h-pyrazol-3-ylamine, 3-amino-1h-pyrazole, 3-aminopyrozole, 5-amino-1h-pyrazole, aminopyrazole, 3-pyrazolamine
74561
1H-pyrazol-5-amine
C1=C(NN=C1)N
This item is not returnable. View return policy
1820-80-0
C3H5N3
83.09
MFCD00005236
JVVRJMXHNUAPHW-UHFFFAOYSA-N
3-aminopyrazole, 1h-pyrazol-3-amine, 1h-pyrazole-5-amine, 5-aminopyrazole, 1h-pyrazol-3-ylamine, 3-amino-1h-pyrazole, 3-aminopyrozole, 5-amino-1h-pyrazole, aminopyrazole, 3-pyrazolamine
74561
1H-pyrazol-5-amine
C1=C(NN=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 33.0°C to 39.0°C
Color White to Yellow
Boiling Point 218.0°C (122.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Beilstein 24, 14
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 83.09
Percent Purity 98%
Physical Form Oily Liquid or Low Melting Crystalline Solid
Chemical Name or Material 3-Aminopyrazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Aminopyrazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'