missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazine-2-carboxamide, 96%, Thermo Scientific Chemicals

Catalog Number p-7055066
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
32587-10-3
C5H6N4O
138.13
MFCD03407446
SFSMATGDLFHTHE-UHFFFAOYSA-N
aminopyrazine carboxamide, 3-aminopyrazinecarboxamide, 3-amino-2-pyrazinecarboxamide, 3-amino-pyrazine-2-carboxylic acid amide, 3-amino-pyrazinecarboxamide, 3-amino-pyrazine-2-carboxamide, 2-pyrazinecarboxamide,3-amino, pyrazinecarboxamide, 3-amino, pyrazinecarboxamide, 3-amino-7ci,9ci
280292
3-aminopyrazine-2-carboxamide
NC(=O)C1=NC=CN=C1N
This item is not returnable. View return policy
32587-10-3
C5H6N4O
138.13
MFCD03407446
SFSMATGDLFHTHE-UHFFFAOYSA-N
aminopyrazine carboxamide, 3-aminopyrazinecarboxamide, 3-amino-2-pyrazinecarboxamide, 3-amino-pyrazine-2-carboxylic acid amide, 3-amino-pyrazinecarboxamide, 3-amino-pyrazine-2-carboxamide, 2-pyrazinecarboxamide,3-amino, pyrazinecarboxamide, 3-amino, pyrazinecarboxamide, 3-amino-7ci,9ci
280292
3-aminopyrazine-2-carboxamide
NC(=O)C1=NC=CN=C1N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 138.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Aminopyrazine-2-carboxamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'