missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltriethoxysilane, TRC

Product Code. 30179248
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179248 50 g 50g
30179226 100 g 100g
30179543 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179248 Supplier Toronto Research Chemicals Supplier No. TRCA62878550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9 H23 N O3 Si
Molecular Weight (g/mol) 221.37
Synonym Propylamine, 3-(triethoxysilyl)- (6CI,8CI), 3-(Triethoxysilyl)-1-propanamine, (3-Aminopropyl)triethoxylsilane, (3-Aminopropyl)triethoxysilane, (Aminopropyl)triethoxysilane, (γ-Aminopropyl)triethoxylsilane, (γ-Aminopropyl)triethoxysilane, 3-(Triethoxysilanyl)propylamine, 3-(Triethoxysilyl)-1-propylamine, 3-(Triethoxysilyl)propanamine, 3-(Triethoxysilyl)propylamine, 3-APTES, A 0750, A 110, A 1100, A 1100 (coupling agent), A 1101, A 1101 coupling agent, A 1102, A 1102 (silane derivative), A 1106, A 1112, A 1112 (coupling agent), A 3648, AGM 9, AMEO, AMS 70, AP 1690, APS, APS (coupling agent), APS-E, APS-E (coupling agent), APTES, APTS, AX 1100, C 50752, CG 550, CY 550, DB 550, DS-AMEO, Deolink Amino TE, Dow Corning Z 6011, Dynasylan 1203, Dynasylan AMEO, Dynasylan AMEO-T, Dynasylan Hydrosil 2627, E-Acroseal, FD 550, FD 550 (coupling agent), GF 93, Geniosil GF 93, H 550, Hydrosil 2627, JH-A 110, KBE 903, KBE 903P, KBE 930, KBP 90, KEM 903, KH 500, KH 507PS10S, KH 550, KH 550 (amine), KS 600, L 6376, LS 3150, LT 550, Momentive A 1100, NB 1114, NSC 95428, NUCA 1100, Nanoflex TTN 500, OFS 6011, PC 1100, Prosil 220, Prosil 221, Pyralin VM 651, S 330, SCA 1100, SCA-A 10M, SH 6011, SIA 0610.0, Sila-Ace 330, Sila-Ace MS 3201, Sila-Ace S 330, Silane A 1100, Silane AMG 9, Silane coupling agent KH550, Silicone A 1100, Silquest 11009, Silquest A 110, Silquest A 1100, Silquest A 1101, Silquest A 1102, Silquest A 1106, Silsoft A 1100, Silwet A 1100, Surfcoat LX, TSC 202, TSL 8331, Toray Silicone Z 6011, Triethoxy(3-aminopropyl)silane, Triethoxy(γ-aminopropyl)silane, U 13, U 13 (coupling agent), Unisil 13, Unisilan 13, VM 651, VM 651 (coupling agent), WD 50, X 12-843, Xiameter OFS 6011, Z 6011, γ-Aminpropyltriethoxysilane, γ-Triethoxysilylpropylamine, 1-Propanamine, 3-(triethoxysilyl)-
IUPAC Name 3-triethoxysilylpropan-1-amine
SMILES CCO[Si](CCCN)(OCC)OCC

Spécification

Quantity 50 g
InChI Formula InChI=1S/C9H23NO3Si/c1-4-11-14(12-5-2,13-6-3)9-7-8-10/h4-10H2,1-3H3
Formula Weight 221.1447
Chemical Name or Material 3-(Triethoxysilyl)-1-propylamine
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.