missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)trimethoxysilane, TRC

Product Code. 30314431
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314431 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314431 Supplier Toronto Research Chemicals Supplier No. TRCA6293205ML
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3-Aminopropyl)trimethoxysilane
CAS 13822-56-5
Molecular Formula C6H17NO3Si
Quantity 5 mL
Synonym 3-(Trimethoxysilyl)-1-propanamine, (gamma-Aminopropyl)trimethoxysilane, 3-(Trimethoxysilanyl)propylamine, 3-(Trimethoxysilyl)-1-propyl Amine3-(Trimethoxysilyl)propylamine, A 0800, A 1001, A 110, A 110 (coupling agent), A 1108, A 1110, A 301, AMMO, APTMS, AY 43-059P, Aminopropyltrimethoxysilane, CG 551, DB 551, Dynasylan AMMO, GX 540, Geniosil GF 96, JH 56, KBM 903, KBM 903P, KBM 903TMI, KH 1101, KH 170, KH 170 (coupling agent), KH 540, KH 551, KH 555, KMB 903, LS 1420, LT 540, NSC 83845, NUC Silane A 1110, PC 1110, S 360, SC 3900, SCA 1103, SCA-A 10E, SIA 0611.0, Si 903, Sila-Ace MS 3301, Sila-Ace S 360, Silane SC 3900, Silquest A 1108, Silquest A 1110, TSL 8330, TSL 8331E, Toray Silicone Z 6610, Trimethoxy(3-aminopropyl)silane, WD 56, Z 6610, gamma-Aminopropyl trimethoxysilane
InChI Formula InChI=1S/C6H17NO3Si/c1-8-11(9-2,10-3)6-4-5-7/h4-7H2,1-3H3
SMILES CO[Si](CCCN)(OC)OC
IUPAC Name 3-trimethoxysilylpropan-1-amine
Molecular Weight (g/mol) 179.29
Formula Weight 179.0978
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.