missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)triethoxysilane, 98%

Product Code. 11463424
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463424 2500 g 2500g
11443424 100 g 100g
11453424 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463424 Supplier Thermo Scientific Alfa Aesar Supplier No. A10668.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alfa Aesar (3-Aminopropyl)triethoxysilane, 98%, Quantity: 100g, Boiling Point: 217 deg.C, CAS: 919-30-2, Molecular Formula: C9H23NO3Si, Molecular Weight: 221.372, Percent Purity: 98%, pH: 11 | CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol

(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.

Solubility
Miscible with toluene, acetone, chloroform and ethanol.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, acids and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9H23NO3Si
Molecular Weight (g/mol) 221.37
MDL Number MFCD00008207,MFCD01324904
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
Synonym 3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
PubChem CID 13521
IUPAC Name 3-triethoxysilylpropan-1-amine
SMILES CCO[Si](CCCN)(OCC)OCC

Specifications

Boiling Point 217°C
pH 11
Quantity 2500 g
Linear Formula H2N(CH2)3Si(OCH2CH3)3
UN Number UN2735
Beilstein 1754988
Flash Point 104°C (219°F)
Solubility Information Miscible with toluene,acetone,chloroform and ethanol.
Formula Weight 221.37
Percent Purity 98%
Odor Amine-like
Refractive Index 1.421
Sensitivity Air and moisture sensitive
Density 0.948
Chemical Name or Material (3-Aminopropyl)triethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (3-Aminopropyl)triethoxysilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: (3-Aminopropyl)triethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.