missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyl Monophosphate, TRC

Product Code. 30011460
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011460 500 mg 500mg
30010587 50 mg 50mg
30006541 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011460 Supplier Toronto Research Chemicals Supplier No. TRCA628750500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-28-9
Molecular Formula C3 H10 N O4 P
Molecular Weight (g/mol) 155.09
Synonym 1-Propanol, 3-amino-, 1-(dihydrogen phosphate), 1-Propanol, 3-amino-, dihydrogen phosphate (ester) (6CI,7CI,8CI,9CI), 3-Aminopropyl dihydrogen phosphate, 3-Aminopropyl monophosphate
IUPAC Name 3-aminopropyl dihydrogen phosphate
SMILES NCCCOP(=O)(O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C3H10NO4P/c4-2-1-3-8-9(5,6)7/h1-4H2,(H2,5,6,7)
Formula Weight 155.0347
Percent Purity >95%
Chemical Name or Material 3-aminopropyl dihydrogen phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.