missing translation for 'onlineSavingsMsg'
Learn More

(3-Aminopropyl)diethoxymethylsilane, 97%

Product Code. 11339155
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339155 100 g 100g
11329155 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339155 Supplier Thermo Scientific Alfa Aesar Supplier No. L16401.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3179-76-8 | C8H21NO2Si | 191.346 g/mol

(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

Solubility
Soluble and reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3179-76-8
Molecular Formula C8H21NO2Si
Molecular Weight (g/mol) 191.346
MDL Number MFCD00039785
InChI Key HXLAEGYMDGUSBD-UHFFFAOYSA-N
Synonym 3-aminopropylmethyldiethoxysilane, 3-diethoxymethylsilyl propylamine, dynasylan 1506, 3-aminopropyl-methyl-diethoxysilane, 1-propanamine, 3-diethoxymethylsilyl, dynasylan 1505, 3-aminopropyl diethoxymethylsilane, 3-diethoxy methyl silyl propan-1-amine, unii-a36to38mqn, gamma-aminopropylmethyldiethoxysilane
PubChem CID 18511
IUPAC Name 3-[diethoxy(methyl)silyl]propan-1-amine
SMILES CCO[Si](C)(CCCN)OCC

Specifications

Boiling Point 197°C to 199°C
Melting Point <-20°C
Quantity 100 g
UN Number UN2735
Beilstein 1744264
Flash Point 85°C (185°F)
Solubility Information Soluble and reacts with water.
Formula Weight 191.35
Percent Purity 97%
Refractive Index 1.427
Sensitivity Air and moisture sensitive
Density 0.92
Chemical Name or Material (3-Aminopropyl)diethoxymethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.